3-(2,3-Dihydrobenzo[b]furan-5-yl)-1H-pyrazole
Lieferant: Apollo Scientific
OR183459-1G
APOSOR183459-1GEA
183
EUR
APOSOR183459-1G
3-(2,3-Dihydrobenzo[b]furan-5-yl)-1H-pyrazole
3-(2,3-Dihydrobenzo[b]furan-5-yl)-1H-pyrazole
Formel:
C₁₁H₁₀N₂O Molecular Weight: 186,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204129 CAS-Nummer: 1005923-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...