Dibenzoylmethan 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Diphenylpropan-1,3-dion, 1,3-Diphenyl-1,3-propandion

OR18346-250G
APOSOR18346-250GEA 37.7 EUR
APOSOR18346-250G
Dibenzoylmethan 98%
Dibenzoylmethan
Formel: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
Molecular Weight: 224,26 g/mol
Siedepunkt: 219…221 °C/18mmHg
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003085
CAS-Nummer: 120-46-7
EINECS: 204-398-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR