3-(2,5-Dichloro-thien-3-yl)-1H-pyrazole

Lieferant: Apollo Scientific

OR183461-1G
APOSOR183461-1GEA 183 EUR
APOSOR183461-1G
3-(2,5-Dichloro-thien-3-yl)-1H-pyrazole
3-(2,5-Dichloro-thien-3-yl)-1H-pyrazole
Formel: C₇H₄Cl₂N₂S
Molecular Weight: 219,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00112160
CAS-Nummer: 166196-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR