3-(Benzo[b]furan-2-yl)-1H-pyrazole

Lieferant: Apollo Scientific

OR183462-1G
APOSOR183462-1GEA 72.5 EUR
APOSOR183462-1G
3-(Benzo[b]furan-2-yl)-1H-pyrazole
3-(Benzo[b]furan-2-yl)-1H-pyrazole
Formel: C₁₁H₈N₂O
Molecular Weight: 184,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00112205
CAS-Nummer: 666728-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR