3-(4-Isobutylphenyl)-1H-pyrazole

Lieferant: Apollo Scientific

OR183466-1G
APOSOR183466-1GEA 183 EUR
APOSOR183466-1G
3-(4-Isobutylphenyl)-1H-pyrazole
3-(4-Isobutylphenyl)-1H-pyrazole
Formel: C₁₃H₁₆N₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08446305
CAS-Nummer: 948293-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR