N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]benzamide

Lieferant: Apollo Scientific

OR183468-1G
APOSOR183468-1GEA 219 EUR
APOSOR183468-1G
N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]benzamide
N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]benzamide
Formel: C₁₈H₁₃N₃O₂
Molecular Weight: 303,315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054361
CAS-Nummer: 1401786-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR