N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]acetamide
Lieferant: Apollo Scientific
OR183469-1G
APOSOR183469-1GEA
219
EUR
APOSOR183469-1G
N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]acetamide
N-[2-(1H-Pyrazol-3-yl)benzofuran-3-yl]acetamide
Formel:
C₁₃H₁₁N₃O₂ Molecular Weight: 241,245 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054367 CAS-Nummer: 1401785-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...