7-Bromo-4-hydroxyquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR183473-1G
APOSOR183473-1GEA 62.5 EUR
APOSOR183473-1G
7-Bromo-4-hydroxyquinoline-3-carboxylic acid
7-Bromo-4-hydroxyquinoline-3-carboxylic acid
Formel: C₁₀H₆BrNO₃
Molecular Weight: 268,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684222
CAS-Nummer: 860205-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR