7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäure

Lieferant: Apollo Scientific

OR183474-1G
APOSOR183474-1GEA 87 EUR
APOSOR183474-1G
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäure
7-Chlor-4-hydroxy-8-methylchinolin-3-carbonsäure
Formel: C₁₁H₈ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02179783
CAS-Nummer: 405923-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR