4-Hydroxy-6-methylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR183475-1G
APOSOR183475-1GEA
94.4
EUR
APOSOR183475-1G
4-Hydroxy-6-methylquinoline-3-carboxylic acid
4-Hydroxy-6-methylquinoline-3-carboxylic acid
Formel:
C₁₁H₉NO₃ Molecular Weight: 203,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00454147 CAS-Nummer: 35973-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...