Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-(tert-butyl)-4-hydroxyquinoline-3-carboxylate

OR183476-250MG
APOSOR183476-250MGEA 21.9 EUR
APOSOR183476-250MG
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate
Formel: C₁₆H₁₉NO₃
Molecular Weight: 273,327 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24843129
CAS-Nummer: 228728-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR