Ethyl 4-hydroxy-8-iodoquinoline-3-carboxylate ≥95%
Lieferant: Apollo Scientific
OR183496-250MG
APOSOR183496-250MGEA
137
EUR
APOSOR183496-250MG
Ethyl 4-hydroxy-8-iodoquinoline-3-carboxylate ≥95%
Ethyl 4-hydroxy-8-iodoquinoline-3-carboxylate
Formel:
C₁₂H₁₀INO₃ Molecular Weight: 343,117 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06808574 CAS-Nummer: 49713-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...