Ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate ≥98%

Lieferant: Apollo Scientific

OR183505-1G
APOSOR183505-1GEA 74.1 EUR
APOSOR183505-1G
Ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate ≥98%
Ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate
Formel: C₁₃H₁₂ClNO₃
Molecular Weight: 265,692 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00173392
CAS-Nummer: 338795-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR