4-Hydroxy-1-indanon

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-HYDROXY-2,3-DIHYDRO-1H-INDEN-1-ONE, 4-hydroxyindan-1-one, 4-Hydroxy-1-indanone

OR18351-1G
APOSOR18351-1GEA 21.9 EUR
APOSOR18351-1G
4-Hydroxy-1-indanon
4-Hydroxyindanone
Formel: C₉H₈O₂
Molecular Weight: 148,161 g/mol
Schmelzpunkt: 242…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143330
CAS-Nummer: 40731-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR