Methyl-5-amino-2-benzo[b]thiophencarboxylat

Lieferant: Apollo Scientific

Synonyme: Methyl 5-aminobenzo[b]thiophene-2-carboxylate

OR183513-1G
APOSOR183513-1GEA 66.7 EUR
APOSOR183513-1G
Methyl-5-amino-2-benzo[b]thiophencarboxylat
Methyl-5-amino-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₉NO₂S
Molecular Weight: 207.25 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04123924
CAS-Nummer: 20699-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR