1-(3-Amino-4-nitrophenyl)piperidin-4-ol

Lieferant: Apollo Scientific

OR183521-1G
APOSOR183521-1GEA 0 EUR
APOSOR183521-1G
1-(3-Amino-4-nitrophenyl)piperidin-4-ol
1-(3-Amino-4-nitrophenyl)piperidin-4-ol
Formel: C₁₁H₁₅N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27950443
CAS-Nummer: 404009-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR