[1-(3-Amino-4-nitrophenyl)piperidin-4-yl]methanol
Lieferant: Apollo Scientific
OR183522-1G
APOSOR183522-1GEA
183
EUR
APOSOR183522-1G
[1-(3-Amino-4-nitrophenyl)piperidin-4-yl]methanol
[1-(3-Amino-4-nitrophenyl)piperidin-4-yl]methanol
Formel:
C₁₂H₁₇N₃O₃ Molecular Weight: 251,282 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1604046-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...