tert-Butyl 4-(3-amino-4-nitrophenyl)piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR183523-1G
APOSOR183523-1GEA 0 EUR
APOSOR183523-1G
tert-Butyl 4-(3-amino-4-nitrophenyl)piperazine-1-carboxylate
tert-Butyl 4-(3-amino-4-nitrophenyl)piperazine-1-carboxylate
Formel: C₁₅H₂₂N₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD12022589
CAS-Nummer: 193902-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR