2-Chlorbenzimidazol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-1H-benzimidazol

OR18361-100G OR18361-25G OR18361-5G
APOSOR18361-100GEA 68.3 EUR
APOSOR18361-100G APOSOR18361-25G APOSOR18361-5G
2-Chlorbenzimidazol 96%
2-Chlorobenzimidazole
Formel: C₇H₅ClN₂
Molecular Weight: 152.58 g/mol
Schmelzpunkt: 180…185 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00051944
CAS-Nummer: 4857-06-1
EINECS: 225-453-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR