2-Aminonicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Aminopyridin-3-carbonsäure, 2-Amino-3-pyridincarbonsäure

OR18366-1G
APOSOR18366-1GEA 21.9 EUR
APOSOR18366-1G
2-Aminonicotinsäure 98%
2-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006318
CAS-Nummer: 5345-47-1
EINECS: 226-296-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR