3-Acetylbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl-m-cyanphenylketon, 3'-Cyanacetophenon
OR18369-25G
APOSOR18369-25GEA
72.5
EUR
APOSOR18369-25G
3-Acetylbenzonitril 98%
3-Acetylbenzonitril
Formel:
C₉H₇NO Molecular Weight: 145.16 g/mol Siedepunkt: 120 Schmelzpunkt: 97…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001806 CAS-Nummer: 6136-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...