Dimethyl-5-nitroisophthalat 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Nitro-1,3-benzoldicarbonsäuredimethylester, 5-Nitroisophthalsäuredimethylester
OR18375-250G
OR18375-100G
OR18375-500G
APOSOR18375-250GEA
37.7
EUR
APOSOR18375-250G
APOSOR18375-100G
APOSOR18375-500G
Dimethyl-5-nitroisophthalat 98+%
Dimethyl-5-nitroisophthalat
Formel:
C₁₀H₉NO₆ Schmelzpunkt: 121…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008429 CAS-Nummer: 13290-96-5 EINECS: 236-307-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...