Cholin-p-toluolsulfonat 85%

Lieferant: Apollo Scientific
Achtung

OR18384-5G OR18384-25G
APOSOR18384-5GEA 36.4 EUR
APOSOR18384-5G APOSOR18384-25G
Cholin-p-toluolsulfonat 85%
Cholin-p-toluolsulfonat
Formel: C₅H₁₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00079051
CAS-Nummer: 55357-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR