Methansulfonamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methylsulphonamide

OR18387-500G OR18387-100G OR18387-50G
APOSOR18387-500GEA 62.5 EUR
APOSOR18387-500G APOSOR18387-100G APOSOR18387-50G
Methansulfonamid 97%
Methansulfonamid
Formel: CH₅NO₂S
Schmelzpunkt: 85…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007940
CAS-Nummer: 3144-09-0
EINECS: 221-553-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR