Methyl-1-methyl-2-sulphanyl-1H-imidazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-mercapto-1-methyl-1H-imidazole-5-carboxylate

OR18399-1G
APOSOR18399-1GEA 42.2 EUR
APOSOR18399-1G
Methyl-1-methyl-2-sulphanyl-1H-imidazole-5-carboxylate
Methyl-1-methyl-2-sulphanyl-1H-imidazole-5-carboxylate
Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00206762
CAS-Nummer: 68892-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR