5-[(2-Methylquinol-4-yl)thio]-1,3,4-thiadiazol-2-amine

Lieferant: Apollo Scientific

Synonyme: 5-((2-Methylquinolin-4-yl)thio)-1,3,4-thiadiazol-2-amine

OR18400-1G
APOSOR18400-1GEA 112 EUR
APOSOR18400-1G
5-[(2-Methylquinol-4-yl)thio]-1,3,4-thiadiazol-2-amine
5-[(2-Methylquinol-4-yl)thio]-1,3,4-thiadiazol-2-amine
Formel: C₁₂H₁₀N₄S₂
Molecular Weight: 274,365 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00105624
CAS-Nummer: 219719-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR