3-Amino-5-phenylthiophene-2-carboxamide

Lieferant: Apollo Scientific
Achtung

OR18408-100MG
APOSOR18408-100MGEA 21.9 EUR
APOSOR18408-100MG
3-Amino-5-phenylthiophene-2-carboxamide
3-Amino-5-phenylthiophene-2-carboxamide
Formel: C₁₁H₁₀N₂OS
Molecular Weight: 218,275 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068129
CAS-Nummer: 122375-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR