2-Methoxy-6-methylbenzaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR18418-1G
APOSOR18418-1GEA
105
EUR
APOSOR18418-1G
2-Methoxy-6-methylbenzaldehyde 97%
2-Methoxy-6-methylbenzaldehyde
Formel:
C₉H₁₀O₂ Molecular Weight: 150,174 g/mol Schmelzpunkt: 38…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235006 CAS-Nummer: 54884-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...