1,3,5-Tris(brommethyl)benzol 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
OR18429-1G
OR18429-5G
OR18429-25G
APOSOR18429-1GEA
21.9
EUR
APOSOR18429-1G
APOSOR18429-5G
APOSOR18429-25G
1,3,5-Tris(brommethyl)benzol 97+%
1,3,5-Tris(brommethyl)benzol
Formel:
C₉H₉Br₃ Molecular Weight: 356.88 g/mol Schmelzpunkt: 94…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00182500 CAS-Nummer: 18226-42-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...