3-Chloro-5-nitroaniline 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR18564-10G
OR18564-1G
OR18564-5G
APOSOR18564-10GEA
0
EUR
APOSOR18564-10G
APOSOR18564-1G
APOSOR18564-5G
3-Chloro-5-nitroaniline 98%
3-Chloro-5-nitroaniline
Formel:
C₆H₅ClN₂O₂ Schmelzpunkt: 131…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01009865 CAS-Nummer: 5344-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...