3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-[2-(3-Formylphenoxy)ethyl]morpholine hydrochloride

OR18570-250MG
APOSOR18570-250MGEA 102 EUR
APOSOR18570-250MG
3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride
3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride
Formel: C₁₃H₁₇NO₃·ClH
Molecular Weight: 271,74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13186601
CAS-Nummer: 1609402-82-5
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR