1,5-Dimethyl-1H-pyrazol-4-boronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1,5-Dimethyl-1H-pyrazol-4-yl)boronic acid

OR18577-100MG OR18577-250MG OR18577-1G
APOSOR18577-100MGEA 77 EUR
APOSOR18577-100MG APOSOR18577-250MG APOSOR18577-1G
1,5-Dimethyl-1H-pyrazol-4-boronsäure 98%
1,5-Dimethyl-1H-pyrazole-4-boronic acid
Formel: C₅H₉BN₂O₂
Molecular Weight: 139.9482 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD13195678
CAS-Nummer: 1204333-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR