1-{[(tert-Butoxy)carbonyl]amino}-4-hydroxycyclohexane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid

OR18581-500MG
APOSOR18581-500MGEA 336 EUR
APOSOR18581-500MG
1-{[(tert-Butoxy)carbonyl]amino}-4-hydroxycyclohexane-1-carboxylic acid
1-{[(tert-Butoxy)carbonyl]amino}-4-hydroxycyclohexane-1-carboxylic acid
Formel: C₁₂H₂₁NO₅
Molecular Weight: 259,3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02683153
CAS-Nummer: 369403-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR