Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate

OR18601-1G
APOSOR18601-1GEA 318 EUR
APOSOR18601-1G
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate
Formel: C₈H₉ClN₂O₂
Molecular Weight: 200,622 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03425963
CAS-Nummer: 188781-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR