1-Methyl-1H-benzo[d]imidazole-2-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR1861-250MG
APOSOR1861-250MGEA
0
EUR
APOSOR1861-250MG
1-Methyl-1H-benzo[d]imidazole-2-carboxylic acid 97%
1-Methyl-1H-benzo[d]imidazole-2-carboxylic acid
Formel:
C₉H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01836869 CAS-Nummer: 20572-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...