3-Acetoxy-4-methoxybenzeneboronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Acetoxy-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate

OR18613-1G OR18613-250MG
APOSOR18613-1GEA 81.2 EUR
APOSOR18613-1G APOSOR18613-250MG
3-Acetoxy-4-methoxybenzeneboronic acid, pinacol ester
3-Acetoxy-4-methoxybenzeneboronic acid, pinacol ester
Formel: C₁₅H₂₁BO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD06795645
CAS-Nummer: 917757-44-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR