3-Nitro-1H-pyrazol

Lieferant: Apollo Scientific
Achtung

OR18620-25G
APOSOR18620-25GEA 21.9 EUR
APOSOR18620-25G
3-Nitro-1H-pyrazol
3-Nitro-1H-pyrazole
Formel: C₃H₃N₃O₂
Molecular Weight: 113,08 g/mol
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00238787
CAS-Nummer: 26621-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR