2-(Cyclohexyloxy)isonicotinic acid

Lieferant: Apollo Scientific

OR18621-1G
APOSOR18621-1GEA 209 EUR
APOSOR18621-1G
2-(Cyclohexyloxy)isonicotinic acid
2-(Cyclohexyloxy)isonicotinic acid
Formel: C₁₂H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09258836
CAS-Nummer: 1019353-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR