5-tert-Butoxycarbonylamino-2-chloro-isonicotinic acid

Lieferant: Apollo Scientific

Synonyme: 5-Amino-2-chloroisonicotinic acid, 5-BOC protected

OR18623-1G
APOSOR18623-1GEA 197 EUR
APOSOR18623-1G
5-tert-Butoxycarbonylamino-2-chloro-isonicotinic acid
5-tert-Butoxycarbonylamino-2-chloro-isonicotinic acid
Formel: C₁₁H₁₃ClN₂O₄
Molecular Weight: 272,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258838
CAS-Nummer: 171178-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR