1-Benzhydryl-3-azetidinol hydrochloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Diphenylmethyl)-3-hydroxyazetidine hydrochloride, 1-Benzhydrylazetidin-3-ol hydrochloride
OR1872-1G
APOSOR1872-1GEA
21.9
EUR
APOSOR1872-1G
1-Benzhydryl-3-azetidinol hydrochloride 95%
1-Benzhydryl-3-azetidinol hydrochloride
Formel:
C₁₆H₁₇NO·HCl Molecular Weight: 275,78 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209447 CAS-Nummer: 90604-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...