Ethyl-4-methyl-1,3-thiazole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR18721-1G OR18721-250MG
APOSOR18721-1GEA 0 EUR
APOSOR18721-1G APOSOR18721-250MG
Ethyl-4-methyl-1,3-thiazole-2-carboxylate
Ethyl-4-methyl-1,3-thiazole-2-carboxylate
Formel: C₇H₉NO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD11112113
CAS-Nummer: 7210-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR