(8S)-8-Amino-7-oxononanoic acid hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Amino-7-oxopelargonic acid hydrochloride
OR1875T-5MG
APOSOR1875T-5MGEA
0
EUR
APOSOR1875T-5MG
(8S)-8-Amino-7-oxononanoic acid hydrochloride
(8S)-8-Amino-7-oxononanoic acid hydrochloride
An intermediate in the synthesis of biotin.
Formel:
C₉H₁₇NO₃·ClH Molecular Weight: 223,697 g/mol Schmelzpunkt: 134…136 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12912236 CAS-Nummer: 177408-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...