2-Bromo-6-methyl-4-nitroanisole

Lieferant: Apollo Scientific
Achtung

OR18801-1G OR18801-5G OR18801-25G
APOSOR18801-1GEA 21.9 EUR
APOSOR18801-1G APOSOR18801-5G APOSOR18801-25G
2-Bromo-6-methyl-4-nitroanisole
2-Bromo-6-methyl-4-nitroanisole
Formel: C₈H₈BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20441781
CAS-Nummer: 1345471-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR