6-Amino-5-chlornicotinonitril

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Amino-3-chloro-5-cyanopyridine

OR18807-1G
APOSOR18807-1GEA 66.7 EUR
APOSOR18807-1G
6-Amino-5-chlornicotinonitril
6-Amino-5-chlornicotinonitril
Formel: C₆H₄ClN₃
Molecular Weight: 153,5691 g/mol
Schmelzpunkt: 188…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03617768
CAS-Nummer: 156361-02-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR