Trisodium naphthalene-1,3,5-trisulphonate ≥96%

Lieferant: Apollo Scientific
Achtung

OR18814-250MG
APOSOR18814-250MGEA 194 EUR
APOSOR18814-250MG
Trisodium naphthalene-1,3,5-trisulphonate ≥96%
Trisodium naphthalene-1,3,5-trisulphonate
Formel: C₁₀H₅O₉S₃·3Na
Molecular Weight: 434,3056 g/mol
MDL: MFCD00065323
CAS-Nummer: 10533-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR