5-Bromo-1,2,3,4-tetrahydroisoquinoline 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR18818-250MG
APOSOR18818-250MGEA
23.2
EUR
APOSOR18818-250MG
5-Bromo-1,2,3,4-tetrahydroisoquinoline 97+%
5-Bromo-1,2,3,4-tetrahydroisoquinoline
Formel:
C₉H₁₀BrN Molecular Weight: 212,0900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08544279 CAS-Nummer: 81237-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...