(2-Carboxyethyl)triphenylphosphoniumbromid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR18825-25G
APOSOR18825-25GEA
71.2
EUR
APOSOR18825-25G
(2-Carboxyethyl)triphenylphosphoniumbromid 97%
(2-Carboxyethyl)triphenylphosphonium bromide
Formel:
C₂₁H₂₀BrO₂P Molecular Weight: 415,2601 g/mol Schmelzpunkt: 198 °C Flammpunkt: >230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031698 CAS-Nummer: 51114-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...