3-Chlor-4-formylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloroisonicotinaldehyde, 3-Chloro-4-pyridinecarboxaldehyde, 3-Chloropyridine-4-carboxaldehyde
OR18827-25G
APOSOR18827-25GEA
120
EUR
APOSOR18827-25G
3-Chlor-4-formylpyridin
3-Chlor-4-formylpyridin
Formel:
C₆H₄ClNO Molecular Weight: 141,56 g/mol Schmelzpunkt: 58…62 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06200712 CAS-Nummer: 72990-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...