(2R)-Acetoxy(phenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-(-)-O-Acetylmandelic acid
OR18839-100G
APOSOR18839-100GEA
75.4
EUR
APOSOR18839-100G
(2R)-Acetoxy(phenyl)essigsäure 98%
(R)-2-Acetoxy-2-phenylacetic acid
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,19 g/mol Schmelzpunkt: 97…99 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004249 CAS-Nummer: 51019-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...