Boc-L-pyroglutamic acid methyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Methyl N-Boc-L-pyroglutamate, Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, Boc-Pyr-OMe, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-Boc-L-pyroglutamic acid methyll ester, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid methyll ester, 1-(tert-Butyl) 2-methyl (S)-5-oxopyrrolidine-1,2-dicarboxylate, Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

OR18848-500G
APOSOR18848-500GEA 92.8 EUR
APOSOR18848-500G
Boc-L-pyroglutamic acid methyl ester
Boc-L-pyroglutamic acid methyl ester
Formel: C₁₁H₁₇NO₅
Molecular Weight: 243.26 g/mol
Schmelzpunkt: 68…74 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06809720
CAS-Nummer: 108963-96-8
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR